2-Thiophenemethanethiol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Mercaptomethyl)thiophene, Thenyl Mercaptan

26511-0010 26511-0050
TS26511-0010EA 178 USD
TS26511-0010 TS26511-0050
2-Thiophenemethanethiol 98%
2-Thiophenemethanethiol
Formula: C₅H₆S₂
MW: 130.22 g/mol
Boiling Pt: 90 °C (15 mmHg)
Density: 1.2 g/cm³
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00107135
CAS Number: 6258-63-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6020 to 1.6040 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR