3-Amino-1H-1,2,4-triazole 95%

Supplier: Thermo Scientific Chemicals
Warning

26457-0100 26457-1000
TS26457-0100EA 32 USD
TS26457-0100 TS26457-1000
3-Amino-1H-1,2,4-triazole 95%
3-Amino-1H-1,2,4-triazole
Formula: C₂H₄N₄
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 149…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00053362
CAS Number: 61-82-5
EINECS: 200-521-5
UN: 3077
ADR: 9,III
Merck Index: 13,00489

Order Now

Specification Test Results

Appearance White to off-white to yellow Powder or flakes
Infrared spectrum Conforms
Melting point 147 to 156 °C
Titration with HClO4 ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR