(±)-Tetrahydro-2-furoic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

26314-0250 26314-1000 26314-5000
TS26314-0250EA 45.9 USD
TS26314-0250 TS26314-1000 TS26314-5000
(±)-Tetrahydro-2-furoic acid 98+%
(±)-Tetrahydro-2-furoic acid
Formula: C₅H₈O₃
Boiling Pt: 128…129 °C (13 mmHg)
Density: 1.17 g/cm³
Flash Pt: 139 °C
Storage Temperature: Ambient
MDL Number: MFCD00022473
CAS Number: 16874-33-2
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4620 (20°C, 589 nm)
GC ≥98.0 %
Titration with NaOH 98.0 to 102.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR