Glycerol formal (=1,3-dioxan-5-ol) mixture of isomers 99+% stabilized

Supplier: Thermo Scientific Chemicals
Warning

25977-0050 25977-2500 25977-5000
TS25977-0050EA 27 USD
TS25977-0050 TS25977-2500 TS25977-5000
Glycerol formal (=1,3-dioxan-5-ol) mixture of isomers 99+% stabilized
Glycerol formal (=1,3-dioxan-5-ol)
Formula: C₄H₈O₃
Boiling Pt: 193…195°C (760 mmHg)
Density: 1.21 g/cm³
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD10039829
CAS Number: 4740-78-7

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4500 to 1.4520 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
Solubility (50 % in water) Clear
GC ≥99.0 % Sum of alpha- and beta-isomers
Color scale ≤20 APHA
pH 4.0 to 6.5 (10 % solution)
Aldehyde (HCHO) ≤200 ppm
Thiodipropionic acid ≤150 ppm
Propyl gallate ≤200 ppm
EDTA-Na ≤200 ppm
Peroxides ≤15 ppm (as H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR