2-Phenyl-5-benzimidazolesulfonic acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ensulizole, Phenylbenzimidazole sulphonic acid, 1H-Benzimidazole-5-sulphonic acid

25970-0050 25970-0250 25970-2500
TS25970-0050EA 29 USD
TS25970-0050 TS25970-0250 TS25970-2500
2-Phenyl-5-benzimidazolesulfonic acid 98%
2-Phenyl-5-benzimidazolesulphonic acid
Formula: C₁₃H₁₀N₂O₃S
MW: 274.29 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00053007
CAS Number: 27503-81-7

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤7 %
Titration excess NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR