(±)-γ-Undecalactone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (±)-Undecan-4-olide, Gamma-undecalactone, Undecanoic gamma-Lactone

25949-0250 25949-1000 25949-5000
TS25949-0250EA 24 USD
TS25949-0250 TS25949-1000 TS25949-5000
(±)-γ-Undecalactone 98%
(±)-γ-Undecalactone
Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
Boiling Pt: 286 °C (760 mmHg)
Density: 0.9425 g/cm³
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00005405
CAS Number: 104-67-6
EINECS: 203-225-4

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4540 (20°C, 589 nm)
GC ≥97.5 %
Specific gravity (25°C) 0.939 to 0.948

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR