2-(2-(Piperazin-1-yl)ethoxy)ethanol 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-[2-(2-Hydroxyethoxy)ethyl]piperazine, 1-Hydroxyethylethoxypiperazine

25911-0050 25911-0250 25911-1000
TS25911-0050EA 45 USD
TS25911-0050 TS25911-0250 TS25911-1000
2-(2-(Piperazin-1-yl)ethoxy)ethanol 95%
2-(2-(Piperazin-1-yl)ethoxy)ethanol
Formula: C₈H₁₈N₂O₂
MW: 174.2407 g/mol
Boiling Pt: 150 °C (13 mmHg)
Density: 1.01 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00074931
CAS Number: 13349-82-1

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Viscous liquid
Infrared spectrum Conforms
Titration Tertiary base ≥94 %
GC ≥94 %
Titration with HClO4 ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR