Methyl-6-methylnicotinate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 6-methylnicotinate

24821-0020 24821-0100
TS24821-0020EA 59 USD
TS24821-0020 TS24821-0100
Methyl-6-methylnicotinate 97%
Methyl-6-methylnicotinate
Formula: C₈H₉NO₂
Boiling Pt: 108…110 °C (22.5 mmHg)
Melting Pt: 31…37 °C
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00006340
CAS Number: 5470-70-2

Order Now

Specification Test Results

Appearance Brown Low melting solid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR