2-Hydroxypyridine-N-oxide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Pyridinol-1-oxide

24379-0250 24379-1000 24379-5000
TS24379-0250EA 89 USD
TS24379-0250 TS24379-1000 TS24379-5000
2-Hydroxypyridine-N-oxide 97%
2-Hydroxypyridine-N-oxide
Formula: C₅H₅NO₂
Melting Pt: 147…152 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00006195
CAS Number: 13161-30-3

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
Melting point 147 to 152 °C
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR