5-Bromo-2-thenoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Bromothiophene-2-carboxylic acid, 5-Bromo-2-thiophenecarboxylic acid
24364-0010
24364-0050
TS24364-0010EA
32
USD
TS24364-0010
TS24364-0050
5-Bromo-2-thenoic acid 97%
5-Bromo-2-thenoic acid
Formula:
C₅H₃BrO₂S Melting Pt: 140…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00079725 CAS Number: 7311-63-9 |
Specification Test Results
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 141 to 145 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...