3-Thenoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Thiophenecarboxylic acid, Thiophene-3-carboxylic acid

24311-0010 24311-0050
TS24311-0010EA 31 USD
TS24311-0010 TS24311-0050
3-Thenoic acid 99%
3-Thenoic acid
Formula: C₅H₄O₂S
MW: 128.15 g/mol
Melting Pt: 137…139 °C
MDL Number: MFCD00005467
CAS Number: 88-13-1
EINECS: 201-802-5
Merck Index: 12,09415

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 137 to 141 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR