Ethyl-(S)-(+)-2-pyrrolidinone-5-carboxylate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl (S)-5-oxopyrrolidine-2-carboxylate, H-Pyr-OEt, Ethyl (S)-(+)-2-pyrrolidone-5-carboxylate

23332-0100 23332-0500
TS23332-0100EA 141 USD
TS23332-0100 TS23332-0500
Ethyl-(S)-(+)-2-pyrrolidinone-5-carboxylate 99%
Ethyl-(S)-(+)-2-pyrrolidinone-5-carboxylate
Formula: C₇H₁₁NO₃
Boiling Pt: 176 °C (12 mmHg)
Melting Pt: 47…51 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064497
CAS Number: 7149-65-7

Order Now

Specification Test Results

Appearance White to cream Low melting solid
Infrared spectrum Conforms
GC ≥98.5 %
Specific optical rotation -2° to -5° (20°C, 589 nm) (c=5, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR