Benzofuran-2-carboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
23326-0010
23326-0050
23326-0250
TS23326-0010EA
40
USD
TS23326-0010
TS23326-0050
TS23326-0250
Benzofuran-2-carboxylic acid 98%
Benzofuran-2-carboxylic acid
Formula:
C₉H₆O₃ Melting Pt: 195…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005848 CAS Number: 496-41-3 EINECS: 207-818-9 |
Specification Test Results
Appearance | White to yellow Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 192 to 196 °C |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...