2-Thienylacetyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Thiopheneacetyl chloride, Thiophene-2-acetyl chloride

23223-0050 23223-0500
TS23223-0050EA 26 USD
TS23223-0050 TS23223-0500
2-Thienylacetyl chloride 99%
2-Thienylacetyl chloride
Formula: C₆H₅ClOS
MW: 160.62
Boiling Pt: 130…135 °C (90 mmHg)
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005456
CAS Number: 39098-97-0
EINECS: 254-290-0
UN: 1760
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear brown Liquid
Infrared spectrum Conforms
Refractive index 1.5500 to 1.5520 (20°C, 589 nm)
GC ≥98.5 %
Free acid ≤2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR