Trioxsalen 97.5%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4,5',8-Trimethylpsoralen, Trisoralen, TMP, 2,5,9-Trimethylfuro[3,2-g]benzopyran-7-one

22988-0010 22988-1000
TS22988-0010EA 495 USD
TS22988-0010 TS22988-1000
Trioxsalen 97.5%
Trioxsalen
Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Melting Pt: 230…234 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005010
CAS Number: 3902-71-4
UN: 1759
ADR: 8,I

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 230 to 237 °C
Loss on drying ≤0.5 % (105 °C, 6 hrs)
HPLC ≥96.5 % (on dried substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR