5-Chloro-2-benzoxazolinone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Chloro-2(3H)-benzoxazolone, 5-Chlorobenzo[d]oxazol-2(3H)-one, Chlorzoxazone, Chloroxazone

22895-0250 22895-1000
TS22895-0250EA 76 USD
TS22895-0250 TS22895-1000
5-Chloro-2-benzoxazolinone 98%
5-Chloro-2-benzoxazolinone
Formula: C₇H₄ClNO₂
MW: 169.57 g/mol
Melting Pt: 189…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00005717
CAS Number: 95-25-0
EINECS: 202-403-9

Order Now

Specification Test Results

Appearance White to yellow-brown Powder
Infrared spectrum Conforms
Melting point 189 to 194 °C
Water ≤1 % (K.F.)
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR