6-Mercaptopurine monohydrate 99.5+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Purinethiol monohydrate, 6-Mercaptopurine hydrate, 7H-Purine-6-thiol hydrate, 6-Thiohypoxanthine monohydrate

22652-0050 22652-0250
TS22652-0050EA 90 USD
TS22652-0050 TS22652-0250
6-Mercaptopurine monohydrate 99.5+%
6-Mercaptopurine monohydrate
Formula: C₅H₄N₄S·H₂O
MW: 170.19 g/mol (152.17 anhy)
Melting Pt: 313 °C
Storage Temperature: Ambient
MDL Number: MFCD03854445
CAS Number: 6112-76-1

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Water 9 to 13 % (K.F.)
HPLC ≥99.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR