2,6-Di-tert-butyl-4-methylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
22452-0050
22452-0250
TS22452-0050EA
143
USD
TS22452-0050
TS22452-0250
2,6-Di-tert-butyl-4-methylpyridine 98%
2,6-Di-tert-butyl-4-methylpyridine
Formula:
C₁₄H₂₃N Melting Pt: 33…36 °C Flash Pt: 83 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006305 CAS Number: 38222-83-2 EINECS: 253-834-4 UN: 1325 |
Specification Test Results
Appearance | Colorless to yellow to orange to light brown Liquid or low melting solid |
Infrared spectrum | Conforms |
Refractive index | 1.4760 to 1.4780 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...