Quinoline 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
22114-0025
22114-0050
22114-1000
22114-5000
TS22114-0025EA
645
USD
TS22114-0025
TS22114-0050
TS22114-1000
TS22114-5000
Quinoline 99%
Quinoline
Formula:
C₉H₇N MW: 129.16 g/mol Boiling Pt: 237 °C (760 mmHg) Melting Pt: –15 °C Density: 1.095 g/cm³ Flash Pt: 101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006736 CAS Number: 91-22-5 EINECS: 202-051-6 UN: 2656 ADR: 6.1,III Merck Index: 13,08160 |
Specification Test Results
Appearance | Clear colorless to dark brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6230 to 1.6270 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...