4-Amino-2-mercaptopyrimidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-2-mercaptopyrimidine, 4-Aminopyrimidine-2-thiol, 4-Amino-2-sulphanylpyrimidine

21477-0010 21477-0050
TS21477-0010EA 62 USD
TS21477-0010 TS21477-0050
4-Amino-2-mercaptopyrimidine 97%
4-Amino-2-mercaptopyrimidine
Formula: C₄H₅N₃S
MW: 127.1676 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006075
CAS Number: 333-49-3

Order Now

Specification Test Results

Appearance Yellow to brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR