3-Aminoquinoline 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
21378-0010
21378-0050
TS21378-0010EA
34
USD
TS21378-0010
TS21378-0050
3-Aminoquinoline 99%
3-Aminoquinoline
Formula:
C₉H₈N₂ MW: 144.18 g/mol Boiling Pt: 311 °C (1013 hPa) Melting Pt: 91…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006772 CAS Number: 580-17-6 EINECS: 209-455-1 |
Specification Test Results
Appearance | White to beige to orange-brown Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 92 to 96 °C |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...