5-Bromonicotinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromo-5-pyridine carboxylic acid, 5-Bromopyridine-3-carboxylic acid, 5-Bromo-3-pyridinecarboxylic acid

21139-0100 21139-0500 21139-2500
TS21139-0100EA 24 USD
TS21139-0100 TS21139-0500 TS21139-2500
5-Bromonicotinic acid 98%
5-Bromonicotinic acid
Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Melting Pt: 178…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00009783
CAS Number: 20826-04-4
EINECS: 244-065-5

Order Now

Specification Test Results

Appearance White to yellow-grey to light brown Powder
Infrared spectrum Conforms
Melting point ≥180 °C
HPLC ≥97.5 %
Titration with TBAH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR