N-(Benzyloxycarbonyloxy)succinimide 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
CBZ-OSu (Z-OSu)
21117-0250
21117-1000
21117-5000
TS21117-0250EA
63
USD
TS21117-0250
TS21117-1000
TS21117-5000
N-(Benzyloxycarbonyloxy)succinimide 98%
N-(Benzyloxycarbonyloxy)succinimide
Formula:
C₁₂H₁₁NO₅ MW: 249.22 g/mol Melting Pt: 77…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005513 CAS Number: 13139-17-8 EINECS: 236-075-3 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 77 to 82 °C |
Loss on drying | ≤0.25 % (60 °C, 2 h, 1 mmHg) |
Solubility | (10 % in acetone) Clear colorless to almost colorless |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...