2,4-Dimethylimidazole 90% tech.

Supplier: Thermo Scientific Chemicals
Danger

21032-0050 21032-0250
TS21032-0050EA 78 USD
TS21032-0050 TS21032-0250
2,4-Dimethylimidazole 90% tech.
2,4-Dimethylimidazole
Formula: C₅H₈N₂
MW: 96.13 g/mol
Boiling Pt: 266 °C (733 mmHg)
Melting Pt: 85…87 °C
MDL Number: MFCD00022365
CAS Number: 930-62-1
EINECS: 213-221-4
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Appearance Yellow to orange Viscous liquid or mass
Infrared spectrum Conforms
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR