Methyl orotate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate

20878-0250 20878-1000
TS20878-0250EA 249 USD
TS20878-0250 TS20878-1000
Methyl orotate 98%
Methyl orotate
Formula: C₆H₆N₂O₄
MW: 170.124 g/mol
Melting Pt: 244 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010564
CAS Number: 6153-44-2

Order Now

Specification Test Results

Appearance Off-white to yellow to light brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR