2-Chloro-3,5-dinitropyridine 98+%

Supplier: Thermo Scientific Chemicals
Danger

20819-0010 20819-0050 20819-0250
TS20819-0010EA 41 USD
TS20819-0010 TS20819-0050 TS20819-0250
2-Chloro-3,5-dinitropyridine 98+%
2-Chloro-3,5-dinitropyridine
Formula: C₅H₂ClN₃O₄
MW: 203.54 g/mol
Melting Pt: 63…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00006233
CAS Number: 2578-45-2
EINECS: 219-937-3
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Yellow Powder or crystals
Infrared spectrum Conforms
Melting point 61 to 66 °C
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR