Papaverine hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Danger

20722-0050 20722-0250 20722-1000
TS20722-0050EA 48 USD
TS20722-0050 TS20722-0250 TS20722-1000
Papaverine hydrochloride 99%
Papaverine hydrochloride
Formula: C₂₀H₂₁NO₄·HCl
MW: 375.85 g/mol
Melting Pt: 226 °C
Storage Temperature: Ambient
MDL Number: MFCD00012745
CAS Number: 61-25-6
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR