2,6-Di-tert-butylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

20464-0050 20464-0250
TS20464-0050EA 181 USD
TS20464-0050 TS20464-0250
2,6-Di-tert-butylpyridine 97%
2,6-Di-tert-butylpyridine
Formula: C₁₃H₂₁N
MW: 191.32 g/mol
Boiling Pt: 209
Melting Pt: 2…4 °C
Density: 0.852 g/cm³
Flash Pt: 72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006306
CAS Number: 585-48-8
EINECS: 209-557-6
Merck Index: 12,03086

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4740 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR