5-Bromopyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning

20430-0250 20430-1000
TS20430-0250EA 74 USD
TS20430-0250 TS20430-1000
5-Bromopyrimidine 98%
5-Bromopyrimidine
Formula: C₄H₃BrN₂
MW: 158.99 g/mol
Boiling Pt: 168 °C (727 mmHg)
Melting Pt: 69…73 °C
MDL Number: MFCD00006117
CAS Number: 4595-59-9
EINECS: 224-992-1

Order Now

Specification Test Results

Appearance Yellow to beige to light brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 69 to 73 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR