4-Nitroquinoline-N-oxide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
20379-0010
20379-0050
20379-2500
TS20379-0010EA
118
USD
TS20379-0010
TS20379-0050
TS20379-2500
4-Nitroquinoline-N-oxide 98%
4-Nitroquinoline-N-oxide
Formula:
C₉H₆N₂O₃ MW: 190.1555 g/mol Melting Pt: 154…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006738 CAS Number: 56-57-5 EINECS: 200-281-1 |
Specification Test Results
Appearance | Ochre to brown Crystalline solid |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...