Tetrahydro-4H-pyran-4-one 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dihydro-2H-pyran-4(3H)-one, Tetrahydro-α-pyran-4-one, Tetrahydro-2H-pyran-4-one, Tetrahydro-γ-pyran-4-one

18944-0050 18944-0250
TS18944-0050EA 120 USD
TS18944-0050 TS18944-0250
Tetrahydro-4H-pyran-4-one 99%
Tetrahydro-4H-pyran-4-one
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 166 °C (760 mmHg)
Density: 1.08 g/cm³
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00006581
CAS Number: 29943-42-8
EINECS: 249-967-2
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4510 to 1.4530 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR