5,6,7,8-Tetrahydroquinoline 98%

Supplier: Thermo Scientific Chemicals
Warning

18887-0050 18887-0250 18887-1000
TS18887-0050EA 45 USD
TS18887-0050 TS18887-0250 TS18887-1000
5,6,7,8-Tetrahydroquinoline 98%
5,6,7,8-Tetrahydroquinoline
Formula: C₉H₁₁N
MW: 133.19 g/mol
Boiling Pt: 218…222 °C (760 mmHg)
Density: 1.03 g/cm³
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00006734
CAS Number: 10500-57-9

Order Now

Specification Test Results

Appearance Colorless to light brown Liquid
Infrared spectrum Conforms
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR