2-Thienylacetic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Thiopheneacetic acid, Thiophene-2-acetic acid, 2-(Thiophen-2-yl)acetic acid

18773-0050 18773-0250 18773-1000
TS18773-0050EA 22 USD
TS18773-0050 TS18773-0250 TS18773-1000
2-Thienylacetic acid 98%
2-Thienylacetic acid
Formula: C₆H₆O₂S
MW: 142.18 g/mol
Boiling Pt: 160 °C (22 mmHg)
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00005458
CAS Number: 1918-77-0
EINECS: 217-639-8
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance Brown Crystalline powder
Infrared spectrum Conforms
Melting point 61 to 65 °C
Titration with NaOH ≥97.5 %
GC ≥88 %
Specific optical rotation +13.0° to +17.0° (20 °C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR