4-Amino-1-benzylpiperidine 98%

Supplier: Thermo Scientific Chemicals
Danger

18766-0050 18766-0250 18766-1000
TS18766-0050EA 35 USD
TS18766-0050 TS18766-0250 TS18766-1000
4-Amino-1-benzylpiperidine 98%
4-Amino-1-benzylpiperidine
Formula: C₁₂H₁₈N₂
Boiling Pt: 152 °C (9.7 mmHg)
Density: 0.93 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006504
CAS Number: 50541-93-0
EINECS: 256-620-9
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5410 to 1.5430 (20°C, 589 nm)
GC ≥97.5 %
Titration with HClO4 ≥97.5 %
Sulfite (SO3) ≤1 ppm
Sulfate (SO4) ≤1 ppm
Free chlorine ≤1 ppm (as Cl)
Ammonium (NH4) ≤3 ppm
Arsenic (As) ≤0.01 ppm
Iron (Fe) ≤0.2 ppm
Heavy metals (as Pb) ≤1 ppm
Extractable organic subst ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR