6-Methoxytryptamine 99%

Supplier: Thermo Scientific Chemicals
Danger

18745-0010 18745-2500
TS18745-0010EA 374 USD
TS18745-0010 TS18745-2500
6-Methoxytryptamine 99%
6-Methoxytryptamine
Formula: C₁₁H₁₄N₂O
Melting Pt: 143…146 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005663
CAS Number: 3610-36-4
EINECS: 222-778-2
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
Melting point 143 to 146 °C
Water ≤1 % (K.F.)
Appearance of solution (2% soln. in ethanol) practically clear, almost colorless to yellowish
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR