Benzimidazole 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1H-Benzo[d]imidazole, 1H-Benzimidazole

18642-0025 18642-0250 18642-1000 18642-5000
TS18642-0025EA 722 USD
TS18642-0025 TS18642-0250 TS18642-1000 TS18642-5000
Benzimidazole 98%
Benzimidazole
Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 360 °C
Melting Pt: 171…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00005585
CAS Number: 51-17-2
EINECS: 200-081-4
Merck Index: 13,01082

Order Now

Specification Test Results

Appearance Beige to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 171 to 174 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR