1-Methylindole 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Methylindole, 1-Methyl-1H-indole

18630-0050 18630-0250 18630-1000
TS18630-0050EA 32 USD
TS18630-0050 TS18630-0250 TS18630-1000
1-Methylindole 98+%
1-Methylindole
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 236…239 °C (760 mmHg)
Melting Pt: 94…95 °C
Density: 1.02 g/cm³
Flash Pt: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005800
CAS Number: 603-76-9
EINECS: 210-057-5
UN: 3334
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6050 to 1.6080 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR