5-Nitro-1,10-phenanthroline 98%

Supplier: Thermo Scientific Chemicals
Warning

18608-0010 18608-0050
TS18608-0010EA 159 USD
TS18608-0010 TS18608-0050
5-Nitro-1,10-phenanthroline 98%
5-Nitro-1,10-phenanthroline
Formula: C₁₂H₇N₃O₂
Melting Pt: 199…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00004981
CAS Number: 4199-88-6
EINECS: 224-097-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow to yellow-brown Crystalline powder
Infrared spectrum Conforms
Melting point 199 to 203 °C
HPLC ≥97.5 %
Optical rotation -65° to -69° (1 % in MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR