6-Amino-1,4-benzodioxane 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,3-Dihydrobenzo[b][1,4]dioxin-6-amine, 1,4-Benzodioxan-6-amine
18578-0050
18578-0500
18578-2500
TS18578-0050EA
38
USD
TS18578-0050
TS18578-0500
TS18578-2500
6-Amino-1,4-benzodioxane 99%
6-Amino-1,4-benzodioxane
Formula:
C₈H₉NO₂ MW: 151.17 g/mol Density: 1.23 g/cm³ Flash Pt: 120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006824 CAS Number: 22013-33-8 EINECS: 244-718-4 UN: 2811 ADR: 6.1 III |
Specification Test Results
Appearance | Yellow to dark brown Solid or liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5970 to 1.6010 (20°C, 589 nm) |
Titration with HClO4 | 94.0 to 106.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...