2-(4-Methylthiazol-5-yl)ethanol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methyl-5-(2-hydroxyethyl)thiazole, 4-Methyl-5-thiazoleethanol, 5-(2-Hydroxyethyl)-4-methylthiazole

18406-0250 18406-1000
TS18406-0250EA 28 USD
TS18406-0250 TS18406-1000
2-(4-Methylthiazol-5-yl)ethanol 98%
2-(4-Methylthiazol-5-yl)ethanol
Formula: C₆H₉NOS
Boiling Pt: 135 °C (7 mmHg)
Density: 1.19 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00005339
CAS Number: 137-00-8
EINECS: 205-272-6
Merck Index: 12,06208

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5480 to 1.5510 (20°C, 589 nm)
Water ≤1 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR