2-Mercaptopyridine-N-oxide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Mercaptopyridine 1-oxide
18267-0010
18267-0050
18267-0250
TS18267-0010EA
35
USD
TS18267-0010
TS18267-0050
TS18267-0250
2-Mercaptopyridine-N-oxide 99%
2-Mercaptopyridine N-oxide
Formula:
C₅H₅NOS MW: 127.17 g/mol Melting Pt: 70…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006196 CAS Number: 1121-31-9 EINECS: 214-329-4 |
Specification Test Results
Appearance | Off-white to yellow to yellow-green to beige to grey-green Powder or crystals or chunks |
Infrared spectrum | Conforms |
Melting point | 68 to 72 °C |
Titration Iodometric | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...