Furan 99+% stabilized

Supplier: Thermo Scientific Chemicals
Danger

18111-0010 18111-0025 18111-2500
TS18111-0010EA 135 USD
TS18111-0010 TS18111-0025 TS18111-2500
Furan 99+% stabilized
Furan
Formula: C₄H₄O
MW: 68.08 g/mol
Boiling Pt: 31.3 °C
Melting Pt: –85.6 °C
Density: 0.936 g/cm³
Flash Pt: –35 °C
Storage Temperature: Ambient
MDL Number: MFCD00003222
CAS Number: 110-00-9
UN: 2389
ADR: 3,I

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4197 to 1.4217 (20°C, 589 nm)
Water ≤0.2 %
GC ≤0.05 % (Furfural)
Stabilizer 0.025 to 0.04 % BHT
Peroxides ≤0.015 % (as H2O2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR