1,4-Dithiane-2,5-diol 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,5-Dihydroxy-1,4-dithiane, p-Dithiane-2,5-diol

17993-0010 17993-2500
TS17993-0010EA 512 USD
TS17993-0010 TS17993-2500
1,4-Dithiane-2,5-diol 96%
1,4-Dithiane-2,5-diol
Formula: C₄H₈O₂S₂
MW: 152.24 g/mol
Melting Pt: 130 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00006659
CAS Number: 40018-26-6
EINECS: 254-751-6
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
GC ≥95.0 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR