2-(Aminomethyl)-1-ethylpyrrolidine 95%

Supplier: Thermo Scientific Chemicals
Danger

17948-0100 17948-0500
TS17948-0100EA 97 USD
TS17948-0100 TS17948-0500
2-(Aminomethyl)-1-ethylpyrrolidine 95%
2-(Aminomethyl)-1-ethylpyrrolidine
Formula: C₇H₁₆N₂
Boiling Pt: 58…60 °C (16 mmHg)
Density: 0.884 g/cm³
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00003178
CAS Number: 26116-12-1
UN: 2734
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4655 to 1.4675 (20°C, 589 nm)
Titration with HClO4 ≥94 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR