5-Methyl-5-phenylhydantoin 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Methyl-5-phenylimidazolidine-2,4-dione

17768-0050 17768-0250
TS17768-0050EA 44 USD
TS17768-0050 TS17768-0250
5-Methyl-5-phenylhydantoin 99%
5-Methyl-5-phenylhydantoin
Formula: C₁₀H₁₀N₂O₂
MW: 190.202 g/mol
Melting Pt: 199…201 °C
Storage Temperature: Ambient
MDL Number: MFCD00005265
CAS Number: 6843-49-8

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 198 to 201 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR