1,4-Dioxa-8-azaspiro[4.5]decane 98%

Supplier: Thermo Scientific Chemicals
Warning

17637-0100 17637-0500
TS17637-0100EA 52 USD
TS17637-0100 TS17637-0500
1,4-Dioxa-8-azaspiro[4.5]decane 98%
1,4-Dioxa-8-azaspiro[4.5]decane
Formula: C₇H₁₃NO₂
Boiling Pt: 108…110 °C (26 mmHg)
Density: 1.11 g/cm³
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00005976
CAS Number: 177-11-7

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4809 to 1.4829 (20°C, 589 nm)
GC ≥97.5 %
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR