2,5-Dimethylfuran 99%

Supplier: Thermo Scientific Chemicals
Danger

17603-0250 17603-1000 17603-5000
TS17603-0250EA 55 USD
TS17603-0250 TS17603-1000 TS17603-5000
2,5-Dimethylfuran 99%
2,5-Dimethylfuran
Formula: C₆H₈O
Boiling Pt: 92…94 °C (760 mmHg)
Melting Pt: –62 °C
Density: 0.9 g/cm³
Flash Pt: –1 °C
Storage Temperature: Ambient
MDL Number: MFCD00003250
CAS Number: 625-86-5
EINECS: 210-914-3
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to amber Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4440 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤20 APHA
Titration RCOCl ≥99.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR