3-Quinolinecarboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

17570-0010 17570-0050 17570-0250
TS17570-0010EA 44 USD
TS17570-0010 TS17570-0050 TS17570-0250
3-Quinolinecarboxylic acid 98%
3-Quinolinecarboxylic acid
Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Melting Pt: 277…283 °C
Storage Temperature: Ambient
MDL Number: MFCD00006770
CAS Number: 6480-68-8
EINECS: 229-337-3

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 277 to 283 °C
HPLC ≥97.5 %
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR