2-Aminobenzimidazole 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Benzo[d]imidazol-2-amine, Benzimidazol-2-ylamine, 2-Amino-1H-benzimidazole

17402-0250 17402-1000
TS17402-0250EA 107 USD
TS17402-0250 TS17402-1000
2-Aminobenzimidazole 99+%
2-Aminobenzimidazole
Formula: C₇H₇N₃
MW: 133.15 g/mol
Boiling Pt: 381 °C (1013 hPa)
Melting Pt: 228…232 °C
MDL Number: MFCD00005596
CAS Number: 934-32-7
EINECS: 213-280-6

Order Now

Specification Test Results

Appearance Yellow to beige to light brown Crystalline powder or crystals or flakes (May darken on storage)
Infrared spectrum Conforms
Titration with HClO4 ≥99.0 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR