3-Furoic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Furan-3-carboxylic acid, 3-Furancarboxylic acid

17245-0050 17245-0250
TS17245-0050EA 93 USD
TS17245-0050 TS17245-0250
3-Furoic acid 99%
3-Furoic acid
Formula: C₅H₄O₃
MW: 112.08 g/mol
Melting Pt: 121…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00005350
CAS Number: 488-93-7
EINECS: 207-689-9

Order Now

Specification Test Results

Appearance Beige Crystalline powder
Infrared spectrum Conforms
Melting point 121 to 124 °C
GC ≥98.5 %
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR